Learn More
Electron Microscopy Sciences Easy-Molds™ for size 00 Capsules, 8 mm diameter
SDP
Catalog No. 5025704
Encompass_Preferred
A newly designed mold which is convenient and offers better embedments. This tray-and-mold in one unit offers a unique specimen block that no other mold can. Each tray contains 2x10 block molds and measures 87x66 mm. Size 8mm diameter corresponds to capsule size "00" and forms a block face of 3mm.
Catalog No. 50-257-04 Supplier Electron Microscopy Sciences Supplier No. 6993105
A newly designed mold which is convenient and offers better embedments. This tray-and-mold in one unit offers a unique specimen block that no other mold can. Each tray contains 2x10 block molds and measures 87x66 mm. Size 8mm diameter corresponds to capsule size "00" and forms a block face of 3mm.