Learn More
Electron Microscopy Sciences Easy-Molds™ for size 3 Capsules, 5.6 mm diameter
SDP
Catalog No. 5025702
Encompass_Preferred
A newly designed mold which is convenient and offers better embedments. This tray-and-mold in one unit offers a unique specimen block that no other mold can. Each tray contains 2x10 block molds and measures 87x66 mm. Size: 5.6mm diameter corresponds to capsules size 3, and forms a block face of 1.6mm.
Catalog No. 50-257-02 Supplier Electron Microscopy Sciences Supplier No. 6993005
A newly designed mold which is convenient and offers better embedments. This tray-and-mold in one unit offers a unique specimen block that no other mold can. Each tray contains 2x10 block molds and measures 87x66 mm. Size: 5.6mm diameter corresponds to capsules size 3, and forms a block face of 1.6mm.