Learn More
Marketlab, Inc. Easy Open Specimen Bags
Description
- Extra-Large 12 in. x 15 in. Size: Accommodates larger specimens and related materials for secure transport and storage.
- Colored Construction: Enhances visibility and supports quick specimen identification.
- Three-Wall Design: Separates documentation from specimens to help maintain organization and reduce contamination risk.
- Micro-Perforated Tear Line: Enables quick and convenient access to contents when needed.
- Secure Zipper Closure: Helps keep contents contained during handling and transport.
- Bold Biohazard Symbol: Provides clear visual identification to support proper handling procedures.
- Healthcare and Laboratory Ready: Designed for specimen collection, transport, and storage in clinical and laboratory environments.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Closure Type | Zipper |
| Color | Clear, Yellow |
| Disposable | Single-use |
| Material | LDPE |
| Sterility | Non-sterile |
| Length (English) | 15 in. |
| Width (English) | 12 in. |
| Product Type | Specimen Bag |
| Length (Metric) | 38 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.