Learn More
Marketlab Easy Open Specimen Bags
Description
- Extra-large color block on front of bag for coding by type or transport requirements
- Three-wall construction with separate document pouch
- Micro-perforated tear line for easy opening
- Zipper closure
- Bold and black SPECIMEN text and biohazard symbol for instant identification
- Preprinted check boxes for clear temperature maintenance instructions
- Bilingual text in English and Spanish
- 6 W x 9 H in.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Closure Type | Zip Closure |
| Color | Blue |
| Disposable | Single-use |
| Material | LDPE |
| Sterility | Non-sterile |
| Width (English) | 6 in. |
| Height (English) | 9 in. |
| Product Type | Specimen Bag |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.