Learn More
Microsolv Technology Corporation Easy Purch Pack Max Rec Autom
Catalog No. NC0731858
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 1.2ml, MRQ, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.
Catalog No. NC0731858 Supplier Microsolv Technology Corporation Supplier No. 9512S1CPT
Vial & Cap Kit Includes 100 EA 1.2ml, MRQ, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.