Learn More
Andwin Scientific Easy Read Ruler
SDP
Catalog No. NC1665151
Encompass
A quick centrifuge of the cell suspension pipetted into a PCV tube reveals a packed cell pellet in the PCV capillary, which can be easily read with the aid of an Easy read ruler.
Catalog No. NC1665151 Supplier Andwin Scientific Supplier No. TP87010
A quick centrifuge of the cell suspension pipetted into a PCV tube reveals a packed cell pellet in the PCV capillary, which can be easily read with the aid of an Easy read ruler.