Learn More
Sigma Aldrich Fine Chemicals Biosciences Easy-ReadR thermometer tem1EA
Catalog No. 502900771
Encompass
Easy-to-read thermometers with black printing on a bright yellow background. filled with contrasting black non-hazardous Enviro-Safe liquid. Calibrated with NIST DKD standards.
Catalog No. 50-290-0771 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z4235131EA
Easy-to-read thermometers with black printing on a bright yellow background. filled with contrasting black non-hazardous Enviro-Safe liquid. Calibrated with NIST DKD standards.