Learn More
Dqe Easy Roller Non-Ambulatory Roller System with Aluminum Pool
SDP
Catalog No. NC1398177
Encompass
The Easy Roller Non-Ambulatory Decon System is designed exclusively for the DQE 4' x 8' Standard Collection Pool (steel or aluminum). These rollers allow the Decon Operations Team to easily decontaminate a non-ambulatory patient. The four independent rollers mount to the pool frame in seconds and assist a backboard to pass over the pool with minimal effort. Available As Easy Roller System with Aluminum-frame Standard Collection Pool (HMK11045A) Easy Roller System with Steel-frame Standard Collection Pool (HMK11045S) Easy Roller Upgrade Kit for existing Standard Collection Pool (HM1045) Specifications Easy Roller System with Collection Pool (HMK11045A), (HM1045) 4 aluminum rollers with a load capacity of 300 lb. Contents One DQE 4' x 8' Standard Collection Pool (Aluminum or Steel Frame) Four rollers Vinyl duffel carry bag
Catalog No. NC1398177 Supplier Dqe Supplier No. HMK11045A
The Easy Roller Non-Ambulatory Decon System is designed exclusively for the DQE 4' x 8' Standard Collection Pool (steel or aluminum). These rollers allow the Decon Operations Team to easily decontaminate a non-ambulatory patient. The four independent rollers mount to the pool frame in seconds and assist a backboard to pass over the pool with minimal effort. Available As Easy Roller System with Aluminum-frame Standard Collection Pool (HMK11045A) Easy Roller System with Steel-frame Standard Collection Pool (HMK11045S) Easy Roller Upgrade Kit for existing Standard Collection Pool (HM1045) Specifications Easy Roller System with Collection Pool (HMK11045A), (HM1045) 4 aluminum rollers with a load capacity of 300 lb. Contents One DQE 4' x 8' Standard Collection Pool (Aluminum or Steel Frame) Four rollers Vinyl duffel carry bag