Learn More
MSC EASY SCRUB EXPS STRTR KIT
Catalog No. 502816446
Encompass
This starter kit is designed to reduce chemical waste and cleaning time compared to traditional mop and bucket methods. It includes a flat mop tool, twelve bottles, a caddy, and two flat mop pads, suitable for various floor cleaning tasks.
  • Lightweight, easy-to-handle tool reduces fatigue
  • Reduces chemical waste and cleaning time
  • Aluminum handle with mop pad holder weighs 1.3 lbs (without solution)
  • Handle adjustable from 39–60 in
  • Mop pads are 10 x 18 in (45.7 cm)
  • Compatible with nearly any brand of cleaning solution
Catalog No. 50-281-6446 Supplier MSC Supplier No. 11564994
This starter kit is designed to reduce chemical waste and cleaning time compared to traditional mop and bucket methods. It includes a flat mop tool, twelve bottles, a caddy, and two flat mop pads, suitable for various floor cleaning tasks.
  • Lightweight, easy-to-handle tool reduces fatigue
  • Reduces chemical waste and cleaning time
  • Aluminum handle with mop pad holder weighs 1.3 lbs (without solution)
  • Handle adjustable from 39–60 in
  • Mop pads are 10 x 18 in (45.7 cm)
  • Compatible with nearly any brand of cleaning solution