Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Easy Scrub Flat Mop Tool With Pad Holder

Catalog No. 17623862 Shop All 3M Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-862 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-862 Supplier 3M™ Supplier No. 7000052514
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Easy Scrub Flat Mop Tool With Pad Holder - Flat mop tool used to apply solution to the floor.

  • A flat wet mop measuring 1.3 in. diameter at it's broadest width and weighing only 1 lb. 3 oz. (without fluid), this commercial mop is ideal for tight spaces such as under toilets, beds and vending machines
  • Makes getting in tight spaces such as under toilets, beds, and vending machines easier
  • Cleaning fluid is contained in the handle and filled from an opening at the top of the handle
  • Pole holds 15 oz. of cleaning chemicals.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Mop
Color Black
Length (English) 16 in.
Quantity 1/Cs.
For Use With (Application) Cleaning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.