Learn More
Description
- A mop pad designed for cleaning uneven surfaces such as ceramic tile, quarry tile or raised rubber flooring
- Available in 18 in. width only
- Can be used with nearly any hook and loop flat mop
- Launderable.
Specifications
Specifications
| Product Type | Microfiber Pad |
| Color | White |
| Length (English) | 18 in. |
| Quantity | 40/Cs. |
| For Use With (Application) | Mopping |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.