Learn More
STEMCELL Technologies EasySep™ Mouse CD11c Positive Selection Kit II, For processing: 2 x 10^9 cells
SDP
Catalog No. NC0802723
Encompass
The EasySep™ Mouse CD11c Positive Selection Kit II isolates highly purified CD11c+ cells from splenocytes or other tissues by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and isolated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated cells are immediately ready for downstream applications such as flow cytometry, culture, and cell-based experiments.This product replaces the EasySep™ Mouse CD11c Positive Selection Kit (Catalog #18758) for even faster cell isolations and does not result in the labeling of isolated cells with PE.
Catalog No. NC0802723 Supplier STEMCELL Technologies Supplier No. 18780
The EasySep™ Mouse CD11c Positive Selection Kit II isolates highly purified CD11c+ cells from splenocytes or other tissues by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and isolated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated cells are immediately ready for downstream applications such as flow cytometry, culture, and cell-based experiments.This product replaces the EasySep™ Mouse CD11c Positive Selection Kit (Catalog #18758) for even faster cell isolations and does not result in the labeling of isolated cells with PE.