Learn More
STEMCELL Technologies EasySep™ Mouse CD4+CD25+ Regulatory T Cell Isolation Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1180697
Encompass
The EasySep™ Mouse CD4+CD25+ Regulatory T Cell Isolation Kit II is designed to isolate CD4+CD25+ T cells from single-cell suspensions of splenocytes or other tissues by negative followed by positive selection. CD4+ T cells are first enriched by negative selection using the EasySep™ Mouse CD4+ T Cell Isolation Cocktail. CD25+ cells are then positively selected from the pre-enriched cells using EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Cocktail.
Catalog No. NC1180697 Supplier STEMCELL Technologies Supplier No. 18783
The EasySep™ Mouse CD4+CD25+ Regulatory T Cell Isolation Kit II is designed to isolate CD4+CD25+ T cells from single-cell suspensions of splenocytes or other tissues by negative followed by positive selection. CD4+ T cells are first enriched by negative selection using the EasySep™ Mouse CD4+ T Cell Isolation Cocktail. CD25+ cells are then positively selected from the pre-enriched cells using EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Cocktail.