Learn More
MSC EASY SHINE FLR FINISH KIT
Catalog No. 5000675767
Encompass
This applicator kit is designed to smoothly apply floor finish and protectors using a backpack-based delivery system. It can reduce labor costs by as much as 70% compared to traditional string mops and buckets. The system is compatible with nearly any brand of floor finish.
  • Smoothly applies floor finish and protectors with a backpack-based delivery system
  • Reduces labor costs by as much as 70% compared to string mops and buckets
  • Compatible with nearly any brand of floor finish
  • Handle is extendable from 43 to 63 in.
Catalog No. 50-006-75767 Supplier MSC Supplier No. 84224534
This applicator kit is designed to smoothly apply floor finish and protectors using a backpack-based delivery system. It can reduce labor costs by as much as 70% compared to traditional string mops and buckets. The system is compatible with nearly any brand of floor finish.
  • Smoothly applies floor finish and protectors with a backpack-based delivery system
  • Reduces labor costs by as much as 70% compared to string mops and buckets
  • Compatible with nearly any brand of floor finish
  • Handle is extendable from 43 to 63 in.