Learn More
Tisch Scientific Easy-to-use and disposable In-Line filters
SDP
Supplier:  Tisch Scientific DF15275
Encompass
Easy-to-use and disposable, these In-Line filters from Tisch contain a hydrophobic PTFE membrane ideal for confining and isolating infectious and hazardous material in vacuum systems and venting applications.
Catalog No. 50-225-1420