Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Flowmeter with Valve, Correlated Reading, Easy-View, 316 SS Housing and Fittings, 150-mm Scale, 155 mL/min Air
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3246414
Encompass
Easy to install and easy to read with 180 viewing. New models with dual-floats allow a higher turndown ratiobetter than 20:1.
Catalog No. NC1664192