Learn More
VAI Easy2Gown™ Mitcool 1700 SMS Classic Cleanroom Coveralls
Catalog No. 19546559 Shop All VAI Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-546-559 Large
19-546-557 Small
19-546-558 Medium
19-546-560 X-Large
19-546-561 2X-Large
19-546-562 3X-Large
19-546-563 4X-Large
19-546-564 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-546-559 Supplier VAI Supplier No. 1800PES1802
Add to Cart
Add to Cart
Breathable Mitcool 1700 coveralls are made of a heavyweight, non-woven, and high-quality MicroPorous coated material. Disposable garments are extremely tough and fluid resistant.
  • Bonded polypropylene with laminate
  • Sewn thumb loops to eliminate shifting
  • Piped seams with reinforced stitches
  • Excellent chemical resistance
  • Disposable
Specifications
Product Type Cleanroom Coveralls
Size Large
Color White
Disposable Single-use
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Hood Type Unhooded
Closure Type Zipper
Seam Type Piped, Stitched
Sterility Sterile
Model 1800P-EHB-S
For Use With (Application) Use in Class 100 (Grade A, ISO 5), Class 10,000 (Grade B, ISO 7) and in Class 100,000 (Grade C, ISO 8) areas in both aseptic and non-aseptic manufacturing and testing operations
Style Classic