Learn More
Med Vet International EasyAngle, J1538, Each
Catalog No. 5000495814
Encompass_Preferred
EasyAngle is an innovative medical device that measures mobility of all relevant joints in the body. EasyAngle is a Swedish innovation and is CE-marked asv a medical device. Motivating the patient With EasyAngle it is easy to measure status and progress in small steps due to the dev ice's high accuracy. Measured angles can easily be communicated and used to motivate patients to follow established rehabilitation programs. Easy to use Measurement with one h and allows for the physiotherapist to support the patient and enables an easy measurement procedure. Only one alignment is needed at a time and the display clearly shows each measured result along with five latest measurements. Accurate EasyAngle has a high precision sensor that is accurate with in ±1 °. Efficient work flow Quick measurement save time. All-in-one EasyAngle can be used for high quality assessment of all relevant joints, replacing the goniometer, inclinometer, CROM-device, BROM-device and scoliometer.
Catalog No. 50-004-95814 Supplier Med Vet International Supplier No. JOR1538
EasyAngle is an innovative medical device that measures mobility of all relevant joints in the body. EasyAngle is a Swedish innovation and is CE-marked asv a medical device. Motivating the patient With EasyAngle it is easy to measure status and progress in small steps due to the dev ice's high accuracy. Measured angles can easily be communicated and used to motivate patients to follow established rehabilitation programs. Easy to use Measurement with one h and allows for the physiotherapist to support the patient and enables an easy measurement procedure. Only one alignment is needed at a time and the display clearly shows each measured result along with five latest measurements. Accurate EasyAngle has a high precision sensor that is accurate with in ±1 °. Efficient work flow Quick measurement save time. All-in-one EasyAngle can be used for high quality assessment of all relevant joints, replacing the goniometer, inclinometer, CROM-device, BROM-device and scoliometer.