Learn More
USA Scientific Inc EasyApp Microplate Film Kit
Catalog No. 50352893
Encompass
EasyApp microplate film kit, includes dispenser and two 100-sheet EasyApp film rolls, -40C to +120C, non-sterile
Catalog No. 50-352-893 Supplier USA Scientific Inc Supplier No. 29785877
EasyApp microplate film kit, includes dispenser and two 100-sheet EasyApp film rolls, -40C to +120C, non-sterile