Learn More
USA Scientific Inc EasyApp Microplate Film Rolls, Sterile
Catalog No. NC0250713
Encompass
EasyApp microplate film 100-sheet rolls, -40C to +120C, 2/pack, sterile
Catalog No. NC0250713 Supplier USA Scientific Inc Supplier No. 29776782
EasyApp microplate film 100-sheet rolls, -40C to +120C, 2/pack, sterile