Learn More
USA Scientific Inc EasyApp microplt film kit
Catalog No. 502745125
Encompass
EasyApp microplate film kit, includes dispenser and two 100-sheet EasyApp film rolls, -40C to +120C, str
Catalog No. 50-274-5125 Supplier USA Scientific Inc Supplier No. 29785887
EasyApp microplate film kit, includes dispenser and two 100-sheet EasyApp film rolls, -40C to +120C, str