Learn More
USA Scientific Inc EasyApp Microporous Film Rolls, Sterile
Catalog No. NC0362966
Encompass
EasyApp microporous film 50-sheet rolls, -20C to +80C, 2/pack, sterile
Catalog No. NC0362966 Supplier USA Scientific Inc Supplier No. 29776282
EasyApp microporous film 50-sheet rolls, -20C to +80C, 2/pack, sterile