Learn More
Berkshire Corporation EasyClean™ 360 Isolator Knitted Polyester Padded Covers
Catalog No. 17010232 Shop All Berkshire Corporation Products
Change view
Click to view available options
Color:
White
Catalog No. Color
17-010-232 White
1 options
Catalog No. 17-010-232 Supplier Berkshire Corporation Supplier No. EC360CHP40
Add to Cart
Add to Cart
The EasyClean™ 360 Isolator Cleaning System includes a light-weight, maneuverable cleaning tool along with knitted polyester covers.
  • Made from Berkshire's Choice™ SuperSorb 100% continuous filament knitted polyester material
  • Laundered and packaged in an ISO Class 4 cleanroom for superior fiber, particle and residue performance
  • Foam-free cushioned design ensures even surface coverage with reduced residues and contaminants
  • Easy installation and secure fit on the mop head
  • Recommended for use with the EasyClean™ 360 Knitted Polyester Elastic Covers
  • Compatible for use in ISO Class 3/EU Grade A environments after sterilizing
Specifications
Product Type Mop Cover
Color Code White
Color White
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Single-use
Color-coded No
Material Polyester