Learn More
Berkshire™ EasyClean™ 360 System
Catalog No. 19266701 Shop All Berkshire Corporation Products
This item is not returnable. View return policy
Catalog No. 19-266-701 Supplier Berkshire™ Supplier No. EC360CHPST40
Add to Cart
Add to Cart
This item is not returnable. View return policy
360 System includes a light-weight, maneuverable cleaning tool along with easy-to-use, laundered polyester knit covers.
360 System includes a light-weight, maneuverable cleaning tool along with easy-to-use, laundered polyester knit covers.
  • Sterile, 100% continuous filament polyester knit covers
  • Cleanroom laundered, knife cut edge
  • Certificate of Sterility with each case and available on the web by lot number 24/7*
*Details: Expiration Date, Radiation Dosage, Lot Information, Meets cGMP Requirements For Traceability.
Specifications
Product Type Choice Isolator Tool Cover
Autoclavable Autoclavable
Color-coded No
Material Polyester Knit (1%)