Learn More
Spherotech EASYCOMP BLANK PART 3 MICRON
Catalog No. NC0838673
Encompass_Preferred
EasyComp Fluorescent Particles, Blank, 1E7/mL, 3.0-3.4 um, 5 mL
Catalog No. NC0838673 Supplier Spherotech Supplier No. ECFPB
EasyComp Fluorescent Particles, Blank, 1E7/mL, 3.0-3.4 um, 5 mL