Learn More
StatLab EasyCut Decal Solution
SDP
Catalog No. 22021438
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
22-021-438 1 gal.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-438 Supplier StatLab Supplier No. DCECDGAL
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
StatLab EasyCut Decal Solution is a medium strength decalcifier that can be used for all routine decalcification needs
  • It is formulated to provide less aggressive decalcification than rapid decalcifying solutions and complete demineralization in a timely manner
  • EasyCut Decal Solution offers an excellent balance between decalcification time versus protection of non-calcium containing tissue elements
Specifications
Product Type Decalcifying Solution
Quantity 1 gal.
Solution Type Decalcifier
Form Solution