Learn More
Ted Pella Inc EasyDip™ Slide Staining Jar, yellow
SDP
Catalog No. NC0261686
Encompass
The staining jar, being made of resistant acetal plastic, will not break like most glass jars do. It will resist to attacks by most staining agents including alcohol and xylene but not phenol. The wide stable base offers greater stability while the inside is recessed, allowing for a smaller reagent volume of only 80ml for 12 slides. Easy to clean and no metals to corrode. Ideal for special stains, frozen sections and special processes. Convenient aluminum stain jar holder can hold 6 containers. Slide Staining Rack and Aluminum 6-Jar Holder sold separately.
Catalog No. NC0261686 Supplier Ted Pella Inc Supplier No. 27324
The staining jar, being made of resistant acetal plastic, will not break like most glass jars do. It will resist to attacks by most staining agents including alcohol and xylene but not phenol. The wide stable base offers greater stability while the inside is recessed, allowing for a smaller reagent volume of only 80ml for 12 slides. Easy to clean and no metals to corrode. Ideal for special stains, frozen sections and special processes. Convenient aluminum stain jar holder can hold 6 containers. Slide Staining Rack and Aluminum 6-Jar Holder sold separately.