Learn More
Ted Pella Inc EasyDip™ Slide Staining Rack
SDP
Catalog No. NC9607106
Encompass
The EasyDip™ Slide Staining Rack will hold up to 12 microscope slides with dimensions such as 25 x 75mm, 3 x 1" and even 26 x 76mm and with a thickness of 1.0 and 1.2mm. The slides fit into individual slots for free passage and rapid drainage of staining fluids. Since they are placed vertically in the rack and not horizontally, their writing area will not be stained by the fluid, allowing their removal without the use of forceps. This staining rack is made of material specially formulated for rapidly drying slides in a microwave processor, or at temperatures up to 121°C (250°F). The lid completely covers the EasyDip™ Slide Staining Jar to minimize spill and evaporation. A handle is permanently attached to the rack for easy insertion and removal of slides without your fingers touching the solution. The base of the rack is placed in the vertical position to secure the slides in place, and is rotated sideways for allowing their easy removal.
Catalog No. NC9607106 Supplier Ted Pella Inc Supplier No. 27325
The EasyDip™ Slide Staining Rack will hold up to 12 microscope slides with dimensions such as 25 x 75mm, 3 x 1" and even 26 x 76mm and with a thickness of 1.0 and 1.2mm. The slides fit into individual slots for free passage and rapid drainage of staining fluids. Since they are placed vertically in the rack and not horizontally, their writing area will not be stained by the fluid, allowing their removal without the use of forceps. This staining rack is made of material specially formulated for rapidly drying slides in a microwave processor, or at temperatures up to 121°C (250°F). The lid completely covers the EasyDip™ Slide Staining Jar to minimize spill and evaporation. A handle is permanently attached to the rack for easy insertion and removal of slides without your fingers touching the solution. The base of the rack is placed in the vertical position to secure the slides in place, and is rotated sideways for allowing their easy removal.