Learn More
STEMCELL Technologies EasyEights™ EasySep™ Magnet, Size: 1 Unit
SDP
Catalog No. NC0651008
Encompass
The EasyEights™ EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when simultaneously processing multiple samples, up to eight samples on each side, a total of 16 samples. The EasyEights™ EasySep™ Magnet has a large sample range, capable of processing small samples, starting at 0.1 x 10^8 cells, to larger samples, up to 8.5 x 10^8 cells. The EasyEights™ EasySep™ Magnet is designed to hold up to 8 standard 5 mL (12 x 75 mm) round-bottom tubes on one side, and 8 standard 14 mL (17 x 95 mm) round-bottom or 15 mL conical tubes on the other side.
Catalog No. NC0651008 Supplier STEMCELL Technologies Supplier No. 18103
The EasyEights™ EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when simultaneously processing multiple samples, up to eight samples on each side, a total of 16 samples. The EasyEights™ EasySep™ Magnet has a large sample range, capable of processing small samples, starting at 0.1 x 10^8 cells, to larger samples, up to 8.5 x 10^8 cells. The EasyEights™ EasySep™ Magnet is designed to hold up to 8 standard 5 mL (12 x 75 mm) round-bottom tubes on one side, and 8 standard 14 mL (17 x 95 mm) round-bottom or 15 mL conical tubes on the other side.