Learn More
Bellco Glass, Inc. EASYFERM+ K8 pH Probe,12x200mm Hamilton
SDP
Catalog No. 501946398
Encompass_Preferred
Dissolved oxygen and PH electrodes are the eyes and ears for many biotechnology processes. High quality sensors are required for critical measurement and Hamilton sensors are some of the fi in the world. Bellco offers Hamilton gel-filled PH and stainless steel polarographic dissolved oxygen sensors with the standard 4 pin T82 K8 and vp style connectors in 120 to 425 mm lengths.
Catalog No. 50-194-6398 Supplier Bellco Glass, Inc. Supplier No. 780821200
Dissolved oxygen and PH electrodes are the eyes and ears for many biotechnology processes. High quality sensors are required for critical measurement and Hamilton sensors are some of the fi in the world. Bellco offers Hamilton gel-filled PH and stainless steel polarographic dissolved oxygen sensors with the standard 4 pin T82 K8 and vp style connectors in 120 to 425 mm lengths.