Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAMILTON PROCESS ANALYTICS EASYFERM PLUS PHI K8 120MM

Catalog No. NC2894803
Encompass

  • pH sensor for demanding applications
  • Accurate and stable pH measurement post-sterilization
  • Pre-pressurized reference system
  • Phermlyte electrolyte minimizes signal drift
  • HP Coatramic diaphragm ensures clog-free operation
  • Measuring range: pH 0 to 14
  • Operating temperature: 0 to 140 °C
  • Pressure range: 0 to 6 bar
  • Autoclavable and CIP compatible
  • Wetted parts include glass, ceramic, and EPDM
  • Sample minimum conductivity: 100 µS/cm
  • ATEX and IECEx approved for hazardous areas

Catalog No. NC2894803 Supplier HAMILTON PROCESS ANALYTICS Supplier No. 2386331513
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • pH sensor for demanding applications
  • Accurate and stable pH measurement post-sterilization
  • Pre-pressurized reference system
  • Phermlyte electrolyte minimizes signal drift
  • HP Coatramic diaphragm ensures clog-free operation
  • Measuring range: pH 0 to 14
  • Operating temperature: 0 to 140 °C
  • Pressure range: 0 to 6 bar
  • Autoclavable and CIP compatible
  • Wetted parts include glass, ceramic, and EPDM
  • Sample minimum conductivity: 100 µS/cm
  • ATEX and IECEx approved for hazardous areas

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.