Learn More
Hamilton Robotics EASYFERM PLUS PHI K8 325
Catalog No. NC2135947
Encompass
Sensor Output- Traditional: mVa-length- 325 mmElectrical Connector-K8.Measuring Range pH 0 to 14.
This item is not returnable. View return policy
Catalog No. NC2135947 Supplier Hamilton Robotics Supplier No. 2386331553
Sensor Output- Traditional: mVa-length- 325 mmElectrical Connector-K8.Measuring Range pH 0 to 14.