Learn More
LUKNOVA INC Easyfil Filter 60Ml, 24/25 Joint Adaptor, Packed With 2.5G Celite, Column Shape, Pe Frit Pore Size 10µm. Integrated Via Welding As One-Piece Leak-Free Filter. Pp Structure Resistant To Common Solvents. Temperature Upto 120C. 25 Pieces Per Pack.
Catalog No. NC0722984
Encompass
Luknova Patented Easyfil Filter. 60Ml Volume, 24/25 Joint Adaptor To Fit Standard Flask And Side-Arm To Connect Vacumm, Packed With 2.5G Celite, Column Shape, Polyethylene Frit Pore Size 10µm. This Unit Comes At 50 Filters Per Pack. Frit, Adaptor, And Funnel Body Were Integrated Via Welded Seals As One-Piece Leak-Free Filter. Polypropylene Structure Resistant To Common Aqueous And Organic Solvents Like Ethyl Acetate , Dimethylformamide (Dmf), Dimethylamine (Dma), Diethyl Ether (Dme), Dichloromethane (Dcm), Tetrahydrofuran (Thf), Hexane, Concentrated Sulfuric Acid (H2So4), Hydrochloric Acid (Hcl), Hydrobromic Acid (Hbr), Hydrogen Iodide (Hi), Acetic Acid, Trifluoroacetic Acid (Tfa), Saturated Naoh And Koh Solution, And Even Dimethyl Sulfoxide (Dmso) At Temperature ≤120C (248F). Safe And Ready To Use For Faster Filtration Without Contamination, Save Time And Solvent, Low-Cost Disposable And Resuable Up To 20 Times. Welded Seals Eliminating Bypass Leakage.
Catalog No. NC0722984 Supplier LUKNOVA INC Supplier No. SX810121
Luknova Patented Easyfil Filter. 60Ml Volume, 24/25 Joint Adaptor To Fit Standard Flask And Side-Arm To Connect Vacumm, Packed With 2.5G Celite, Column Shape, Polyethylene Frit Pore Size 10µm. This Unit Comes At 50 Filters Per Pack. Frit, Adaptor, And Funnel Body Were Integrated Via Welded Seals As One-Piece Leak-Free Filter. Polypropylene Structure Resistant To Common Aqueous And Organic Solvents Like Ethyl Acetate , Dimethylformamide (Dmf), Dimethylamine (Dma), Diethyl Ether (Dme), Dichloromethane (Dcm), Tetrahydrofuran (Thf), Hexane, Concentrated Sulfuric Acid (H2So4), Hydrochloric Acid (Hcl), Hydrobromic Acid (Hbr), Hydrogen Iodide (Hi), Acetic Acid, Trifluoroacetic Acid (Tfa), Saturated Naoh And Koh Solution, And Even Dimethyl Sulfoxide (Dmso) At Temperature ≤120C (248F). Safe And Ready To Use For Faster Filtration Without Contamination, Save Time And Solvent, Low-Cost Disposable And Resuable Up To 20 Times. Welded Seals Eliminating Bypass Leakage.