Learn More
METTLER TOLEDO EASYFIT 22, PROCESS ADAPTOR PG
Supplier:  METTLER TOLEDO 52403952
Encompass
Mettler Toledo:EasyFit 22Process Adaptor PG 13.5 to 3/4" NPT M, for 12 mm probes, in 316L*22.03
Catalog No. NC2110258