Learn More
Weber Scientific EASYGEL POTATO DEXTROSE 50/PK
SDP
Catalog No. NC0210603
Encompass
With Antibiotic, pH balanced (acidified.) Yeast are shiny. Whitemolds have fuzzy surface in various colors. 50 bottles of liquid medium and 50 treated petri dishes.
Catalog No. NC0210603 Supplier Weber Scientific Supplier No. 309340
With Antibiotic, pH balanced (acidified.) Yeast are shiny. Whitemolds have fuzzy surface in various colors. 50 bottles of liquid medium and 50 treated petri dishes.