Learn More
Pipette Supplies, Inc. EASYPET 3 SILICONE ADAPTER
SDP
Catalog No. NC1367279
Encompass
Silicone adapter that fits between the aspirating cone and the filter.International Part # 4420801004EP 022236105Made in Germany
Catalog No. NC1367279 Supplier Pipette Supplies, Inc. Supplier No. EP 022236105
Silicone adapter that fits between the aspirating cone and the filter.International Part # 4420801004EP 022236105Made in Germany