Learn More
Research Products International Corp Easyplate Dish Rack, Natural, 84 Capacity
SDP
Catalog No. 501977394
Encompass
This rack solves the problem of toppling stacks of petri dishes. Cleverly designed elastic grippers hold plates snuggly in place-even while in transit. Plates store inverted to eliminate buildup of condensation. Available in colors to dress up the lab or help identify plates. Constructed of heavy duty polycarbonate for rugged durability. Autoclavable.
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7394 Supplier Research Products International Corp Supplier No. 248854
This rack solves the problem of toppling stacks of petri dishes. Cleverly designed elastic grippers hold plates snuggly in place-even while in transit. Plates store inverted to eliminate buildup of condensation. Available in colors to dress up the lab or help identify plates. Constructed of heavy duty polycarbonate for rugged durability. Autoclavable.
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.