Learn More
Contec EasyReach Sterile Ultrasonically Sealed Polyester Cleaning Pads - Bulk
Catalog No. 17111298 Shop All Contec Inc. Products
Change view
Click to view available options
Autoclavable:
Not Autoclavable
Catalog No. Autoclavable
17-111-298 Not Autoclavable
1 options
Catalog No. 17-111-298 Supplier Contec Supplier No. MEQT0022B
Add to Cart
Add to Cart
Highly absorbent sterile polyester pads ensure even coverage of disinfectant or IPA.
  • Single-use sealed polyester
  • Provide a low level of particulates, which is essential for use in product contact areas
  • Very absorbent, ensuring good even coverage of disinfectant or IPA
  • An easy grab edge on the cleaning pad ensures the pad fits tightly on the mop frame
  • Sterile cleaning pads are presaturated with 70% IPA
Specifications
Product Type Mop Cover
Sterility Sterile
Autoclavable Not Autoclavable
Color-coded No
Material Polyester