Learn More
BURKLE INC EasySampler, V4A, length 41.14 in (104.5 cm), volume 6.76 oz (200 ml)
Catalog No. 5000026096
Encompass_Preferred
The EasySampler offers an innovative solution for fast sampling and emptying of free-flowing bulk materials by allowing the sample to be emptied quickly and directly into the collection container. By pushing down on the handle, the sampler tip opens and the sample can be emptied without turning or disassembling the sampler. The smooth, back-edge-free design ensures complete emptying and prevents buildup of residual material, allowing for quick and repeated sampling. In addition, the sampler can be easily disassembled for thorough cleaning. The use of high quality 316L (1.4404) stainless steel ensures durability, reliability and purity during sampling. The EasySampler is specifically designed for free-flowing or self-flowing materials and greatly simplifies sampling.
Catalog No. 50-000-26096 Supplier BURKLE INC Supplier No. 53601000
Add to Cart
Add to Cart
The EasySampler offers an innovative solution for fast sampling and emptying of free-flowing bulk materials by allowing the sample to be emptied quickly and directly into the collection container. By pushing down on the handle, the sampler tip opens and the sample can be emptied without turning or disassembling the sampler. The smooth, back-edge-free design ensures complete emptying and prevents buildup of residual material, allowing for quick and repeated sampling. In addition, the sampler can be easily disassembled for thorough cleaning. The use of high quality 316L (1.4404) stainless steel ensures durability, reliability and purity during sampling. The EasySampler is specifically designed for free-flowing or self-flowing materials and greatly simplifies sampling.