Learn More
BURKLE INC EasyScoop, aluminium, 67.63 oz. (2000 ml), inner grip
Catalog No. 5000026055
Encompass_Preferred
When sampling bulk goods with a high specific weight, conventional scoops subject the user‘s wrist to great stress. EasyScoop with its centrally arranged inner grip is ergonomically designed and ensures easy and safe work. As EasyScoop is considerably shorter than conventional scoops, it is easier to take samples from narrow containers.
Catalog No. 50-000-26055 Supplier BURKLE INC Supplier No. 53562000
When sampling bulk goods with a high specific weight, conventional scoops subject the user‘s wrist to great stress. EasyScoop with its centrally arranged inner grip is ergonomically designed and ensures easy and safe work. As EasyScoop is considerably shorter than conventional scoops, it is easier to take samples from narrow containers.