Learn More
LAMDA BIOTECH INC EasyScript Plus™ cDNA Synthesis Kit
Catalog No. 502045780
Encompass_Preferred
EasyScript Plus™ cDNA Synthesis Kit is a complete system for efficient synthesis of first strand cDNA from RNA templates with secondary structure and high GC contents. The kit utilizes a special Reverse Transcriptase, the EasyScript Plus™, which is a novel recombinant reverse transcriptase that exhibits much higher efficiency in the first strand cDNA synthesis from RNA templates with secondary structures and high GC contents. The EasyScript PlusTM Reverse Transcriptase is engineered to work under high temperatures (50°C-55°C), which can further facilitate to resolve the secondary structures and high GC problems of RNA. Due to this feature, full-length cDNA can be synthesized from RNA templates that are up to 15 kb.
Catalog No. 50-204-5780 Supplier LAMDA BIOTECH INC Supplier No. G236
EasyScript Plus™ cDNA Synthesis Kit is a complete system for efficient synthesis of first strand cDNA from RNA templates with secondary structure and high GC contents. The kit utilizes a special Reverse Transcriptase, the EasyScript Plus™, which is a novel recombinant reverse transcriptase that exhibits much higher efficiency in the first strand cDNA synthesis from RNA templates with secondary structures and high GC contents. The EasyScript PlusTM Reverse Transcriptase is engineered to work under high temperatures (50°C-55°C), which can further facilitate to resolve the secondary structures and high GC problems of RNA. Due to this feature, full-length cDNA can be synthesized from RNA templates that are up to 15 kb.