Learn More
STEMCELL Technologies EasySep™ Mouse F4/80 Positive Selection Kit, For processing: 2 x 10^9 cells
SDP
Catalog No. NC2010419
Encompass
Easily isolate F4/80+ macrophages in as little as 24 minutes—without the use of columns—from mouse splenocytes, lung tissue, and peritoneal lavage.Using immunomagnetic positive selection, F4/80+ cells are labeled with antibodies and dextran-coated magnetic particles (RapidSpheres™), for column-free cell separation using an EasySep™ magnet. Isolated cells are highly purified and ready for your downstream applications.Visit www.EasySep.com (http://easysep.com) to learn more about column-free EasySep™ cell separation technology.
Catalog No. NC2010419 Supplier STEMCELL Technologies Supplier No. 1000659
Easily isolate F4/80+ macrophages in as little as 24 minutes—without the use of columns—from mouse splenocytes, lung tissue, and peritoneal lavage.Using immunomagnetic positive selection, F4/80+ cells are labeled with antibodies and dextran-coated magnetic particles (RapidSpheres™), for column-free cell separation using an EasySep™ magnet. Isolated cells are highly purified and ready for your downstream applications.Visit www.EasySep.com (http://easysep.com) to learn more about column-free EasySep™ cell separation technology.