Learn More
STEMCELL Technologies EasySep™ Release Mouse Biotin Positive Selection Kit, Size: 1 x 10^9 cells
SDP
Catalog No. NC1465595
Encompass
The EasySep™ Release Mouse Biotin Positive Selection Kit is designed to isolate cells that are labeled with biotin-conjugated antibodies from mouse tissue by immunomagnetic positive selection.Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated, biotin-antibody labeled cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins, or other chemically related ligands.This product replaces the EasySep™ Mouse Biotin Positive Selection Kit (Catalog #18556), providing highly purified particle-free cells.
Catalog No. NC1465595 Supplier STEMCELL Technologies Supplier No. 17655
The EasySep™ Release Mouse Biotin Positive Selection Kit is designed to isolate cells that are labeled with biotin-conjugated antibodies from mouse tissue by immunomagnetic positive selection.Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated, biotin-antibody labeled cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins, or other chemically related ligands.This product replaces the EasySep™ Mouse Biotin Positive Selection Kit (Catalog #18556), providing highly purified particle-free cells.