Learn More
STEMCELL Technologies EasySep™ Direct HLA Crossmatch B Cell Isolation Kit, For processing: 100 mL whole blood processed
SDP
Catalog No. NC1744085
Encompass
The EasySep™ Direct HLA Crossmatch B Cell Isolation Kit can be used to isolate highly purified B cells directly from human whole blood, spleen, or lymph nodes by immunomagnetic negative selection. This cell isolation kit targets non-B cells for removal with antibodies recognizing specific surface markers. Unwanted cells are labeled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet. Desired cells are simply collected into a new tube and and are immediately available for downstream applications such as flow cytometry crossmatch.
Catalog No. NC1744085 Supplier STEMCELL Technologies Supplier No. 19684
The EasySep™ Direct HLA Crossmatch B Cell Isolation Kit can be used to isolate highly purified B cells directly from human whole blood, spleen, or lymph nodes by immunomagnetic negative selection. This cell isolation kit targets non-B cells for removal with antibodies recognizing specific surface markers. Unwanted cells are labeled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet. Desired cells are simply collected into a new tube and and are immediately available for downstream applications such as flow cytometry crossmatch.