Learn More
STEMCELL Technologies EasySep™ Direct Human Basophil Isolation Kit, For processing: 100 mL whole blood processed
SDP
Catalog No. NC1736333
Encompass
The EasySep™ Direct Human Basophil Isolation Kit isolates functional, highly purified basophils directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation, or other processing steps are required and isolated cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications.
Catalog No. NC1736333 Supplier STEMCELL Technologies Supplier No. 19667
The EasySep™ Direct Human Basophil Isolation Kit isolates functional, highly purified basophils directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation, or other processing steps are required and isolated cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications.