Learn More
STEMCELL Technologies EasySep™ Direct Human Monocyte Isolation Kit for RoboSep™, For processing: 100 mL whole blood processed
SDP
Catalog No. 501974707
Encompass
The EasySep™ Direct Human Monocyte Isolation Kit isolates functional, highly purified CD14+ monocytes directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation or other processing steps are required and isolated cells are immediately available for flow cytometry, functional assays, culture and other downstream applications.
Catalog No. 50-197-4707 Supplier STEMCELL Technologies Supplier No. 19669RF
The EasySep™ Direct Human Monocyte Isolation Kit isolates functional, highly purified CD14+ monocytes directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation or other processing steps are required and isolated cells are immediately available for flow cytometry, functional assays, culture and other downstream applications.