Learn More
STEMCELL Technologies EasySep™ Direct Human Monocyte Isolation Kit, For processing: 100 mL whole blood processed
SDP
Catalog No. NC0912499
Encompass
The EasySep™ Direct Human Monocyte Isolation Kit isolates functional, highly purified CD14+ monocytes directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation or other processing steps are required and isolated cells are immediately available for flow cytometry, functional assays, culture and other downstream applications.
Catalog No. NC0912499 Supplier STEMCELL Technologies Supplier No. 19669
The EasySep™ Direct Human Monocyte Isolation Kit isolates functional, highly purified CD14+ monocytes directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation or other processing steps are required and isolated cells are immediately available for flow cytometry, functional assays, culture and other downstream applications.