Learn More
STEMCELL Technologies EasySep™ Direct Human Neutrophil Isolation Kit, For processing: 100 mL whole blood processed
SDP
Catalog No. NC0892435
Encompass
The EasySep™ Direct Human Neutrophil Isolation Kit isolates functional, highly purified neutrophils directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation or other processing steps are required and isolat​ed cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications.This product replaces the EasySep™ Human Neutrophil Isolation Kit (Catalog #19257) for even faster cell isolations.
This item is not returnable. View return policy
Catalog No. NC0892435 Supplier STEMCELL Technologies Supplier No. 19666
The EasySep™ Direct Human Neutrophil Isolation Kit isolates functional, highly purified neutrophils directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation or other processing steps are required and isolat​ed cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications.This product replaces the EasySep™ Human Neutrophil Isolation Kit (Catalog #19257) for even faster cell isolations.