Learn More
STEMCELL Technologies EasySep™ Direct Human Total Lymphocyte Isolation Kit for RoboSep™, For processing: 100 mL whole blood processed
SDP
Catalog No. 501974702
Encompass
The EasySep™ Direct Human Total Lymphocyte Isolation Kit isolates highly purified lymphocytes directly from human whole blood by immunomagnetic negative selection. Isolated cells are untouched and available for direct use in flow cytometry, culture​,​ DNA/RNA extraction or other downstream applications.​
Catalog No. 50-197-4702 Supplier STEMCELL Technologies Supplier No. 19655RF
The EasySep™ Direct Human Total Lymphocyte Isolation Kit isolates highly purified lymphocytes directly from human whole blood by immunomagnetic negative selection. Isolated cells are untouched and available for direct use in flow cytometry, culture​,​ DNA/RNA extraction or other downstream applications.​