Learn More
STEMCELL Technologies EasySep™ Human CD19 Positive Selection Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1476303
Encompass
The EasySep™ Human CD19 Positive Selection Kit II is designed to isolate CD19+ cells from fresh or previously frozen peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is expressed on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.This product replaces the EasySep™ Human CD19 Positive Selection Kit (Catalog #18054) for even faster cell isolations.
Catalog No. NC1476303 Supplier STEMCELL Technologies Supplier No. 17854
The EasySep™ Human CD19 Positive Selection Kit II is designed to isolate CD19+ cells from fresh or previously frozen peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is expressed on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.This product replaces the EasySep™ Human CD19 Positive Selection Kit (Catalog #18054) for even faster cell isolations.